Would you like to log in again?
SensoLyte® NADP/NADPH Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72206
103010-620EA 809.69 USD InStock
103010-620
SensoLyte® NADP/NADPH Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Nicotinamide nucleotides are key players in the energy transforming and oxidation-reduction reactions of a cell
The SensoLyte® NADP/NADPH Assay Kit utilizes an optimized enzymatic recycling method for the quantification of NADP/NADPH without interfering with NAD/NADH. NADP is converted to NADPH in this enzyme-cycling reaction. The kit contains a fluorogenic reagent, producing fluorescence when reacted with NADPH. The resulting red fluorescence can be monitored at excitation /emission= 560/590 nm. The intensity of the fluorescence produced is proportional to NADP/NADPH concentration.
NADP and NADPH act as allosteric effectors in modulating enzyme activities. The reducing power of NADPH is a required cofactor for enzymes that are pro-oxidants, such as nitric oxide synthase and NADPH oxidase. NADP/NADPH ratio is involved in the control of the oxidative shunt. Each of these forms has a particular role during differentiation and similarity processes as well as being target functions in the cytotoxic action of ionizing radiations and alkylating agents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty