Would you like to log in again?
SensoLyte® 520 Factor Xa Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72208
103010-624EA 887.15 USD InStock
103010-624
SensoLyte® 520 Factor Xa Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Factor Xa (FXa) is a serine endopeptidase composed of two disulfide-linked subunits
The SensoLyte® 520 Factor Xa Assay Kit provides a convenient assay for high throughput screening of FXa inhibitors or continuous assay of enzyme activity. It detects FXa activity using a 5-FAM/QXL® 520 fluorescence resonance energy transfer (FRET) peptide. In the intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon cleavage into two separate fragments by FXa, the fluorescence of 5-FAM is recovered, and can be monitored at excitation/emission wavelengths = 490 nm/520 nm. With excellent fluorescence quantum yield and longer wavelength, 5-FAM shows less interference from autofluorescence of test compounds and cellular components. The assays are performed in a convenient 96-well microplate format.
FXa leads to blood clot formation by converting prothrombin to thrombin through the prothrombinase complex. FXa is generated from zymogen Factor X via the intrinsic and extrinsic pathways and is the rate-limiting step in the propagation of thrombin generation. In the presence of Ca2+ ions, FXa forms prothrombinase with factor Va on the phospholipid membrane of the activated platelets. FXa has emerged as an attractive target for drug discovery for thromboembolic diseases.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty