Would you like to log in again?
SensoLyte® 520 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72209
103010-626EA 887.15 USD InStock
103010-626
SensoLyte® 520 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Enterokinase is a heterodimeric serine protease produced by cells in the duodenal wall
The SensoLyte® 520 Enterokinase Assay Kit is optimized for detecting activity of enterokinase. This kit employs a novel internally quenched 5-FAM/QXL FRET substrate for enterokinase. Enterokinase cleaves the FRET substrate into two separate fragments resulting in release of 5-FAM fluorescence which can be monitored at excitation/emission= 490/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. The assay can detect as low as 1.25 ng/mL of active enterokinase.
It converts trypsinogen to trypsin, indirectly activating a number of pancreatic digestive enzymes. Enterokinase is a key enzyme for intestinal digestion of proteins. Its deficiency may cause severe protein malabsorption. Enterokinase also plays an important role in acute experimental and clinical pancreatitis. The high degree of specificity exhibited by enterokinase for cleaving affinity or other tags from recombinant proteins makes it an ideal tool for use in biochemical applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty