Would you like to log in again?
SensoLyte® Rh110 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72210
103010-628EA 809.69 USD InStock
103010-628
SensoLyte® Rh110 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
Enterokinase is a heterodimeric serine protease produced by cells in the duodenal wall
The SensoLyte® Rh110 Enterokinase Assay Kit is optimized for detection of enterokinase activity using a fluorogenic substrate. Upon enzyme cleavage, this substrate generates the Rh110 (rhodamine 110) fluorophore with bright green fluorescence that can be detected at excitation/emission=490/520 nm. The longer-wavelength spectra and higher extinction coefficient of the Rh110 provide greater sensitivity and less interference from other reaction components. The assay can detect as low as 0.31 ng/mL active enterokinase.
It converts trypsinogen to trypsin, indirectly activating a number of pancreatic digestive enzymes. Enterokinase is a key enzyme for intestinal digestion of proteins and its deficiency may cause severe protein malabsorption. Enterokinase also plays an important role in acute experimental and clinical pancreatitis. The high degree of specificity exhibited by enterokinase for cleaving affinity or other tags from recombinant proteins makes it an ideal tool for use in biochemical applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty