Would you like to log in again?
SensoLyte® Thioflavin T β-Amyloid (1-40) Aggregation Kit, AnaSpec
Supplier: AnaSpec
AS-72213
103010-634EA 907.62 USD InStock
103010-634
SensoLyte® Thioflavin T β-Amyloid (1-40) Aggregation Kit, AnaSpec
Assays Enzyme Assays
The SensoLyte® ThT β-Amyloid (1-40) Aggregation kit provides a convenient and standard method to measure Aβ40 aggregation using Thioflavin T dye
Alzheimer's disease (AD), the most common cause of dementia, is characterized by the presence of senile plaques and neurofibrillary tangles, surrounded by damaged neurons. Beta-Amyloid (Aβ) peptides, Aβ40 (1-40) and Aβ42 (1-42), were found to be a major component of the above plaques. Many studies suggest that these peptides can form toxic oligomers and fibrils under physiological conditions and rapidly aggregate. Since Aβ aggregation is evidently an essential event in the pathogenesis of AD, a reliable assay is important to study Aβ fibrillation kinetics and screen for Aβ aggregation inhibitors.
Aβ40 peptide is pretreated to ensure it is in a monomeric state. An optimized fibrillation buffer is included with the kit, and two known inhibitors are supplied as controls. The assay is based on the property of ThT dye in which fluorescence (Ex/Em=440/484 nm) is increased when bound to aggregated Aβ peptides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty