Would you like to log in again?
SensoLyte® 520 Neprilysin Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72223
103010-662EA 887.15 USD InStock
103010-662
SensoLyte® 520 Neprilysin Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Neprilysin (NEP) is a transmembrane metallopeptidase normally expressed by a variety of tissues
The SensoLyte® 520 Neprilysin Assay Kit employs a novel internally quenched 5-FAM/QXL® FRET substrate for the detection of neprilysin activity. The enzyme cleaves the FRET substrate into two separate fragments resulting in the release of 5-FAM fluorescence, which can be monitored at excitation /emission= 490/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. The assay can detect as low as 0.78 ng/mL of active Neprilysin.
It is also known as neutral endopeptidase, and enkephalinase. NEP cleaves peptides at the N-terminal side of hydrophobic amino acid residues and is responsible for the degradation and inactivation of a variety of physiological substrates. NEP is a major extracellular amyloid beta-peptide degrading enzyme in the brain and targeting NEP is considered a potential therapeutic strategy for the prevention and treatment of Alzheimer's disease. NEP has also been implicated in the pathogenesis of hypertension, diabetes, and cancer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty