Would you like to log in again?
SensoLyte® Plus 520 MMP-2 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Supplier: AnaSpec
AS-72224
103010-664EA 969 USD InStock
103010-664
SensoLyte® Plus 520 MMP-2 Assay Kit (Fluorimetric and Enhanced Selectivity), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Plus 520 MMP-2 Assay Kit is designed specifically for detecting MMP-2 activity in biological samples, such as culture medium, serum, plasma, synovial fluid, and tissue homogenate, which may contain multiple MMPs. A monoclonal anti-human-MMP-2 antibody is employed to pull down MMP-2 from the mixture first. MMP-2 activity is then quantified by a 5-FAM/QXL® 520 fluorescence resonance energy transfer (FRET) peptide. The fluorescence signal is monitored at Ex/Em=490 nm/520 nm upon MMP-2-induced cleavage of the FRET substrate. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. Ample materials are provided to perform 96 assays in a 96-well format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty