Would you like to log in again?
SensoLyte® 520 BACE2 Activity Assay (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72225
103010-666EA 809.69 USD InStock
103010-666
SensoLyte® 520 BACE2 Activity Assay (Fluorimetric), AnaSpec
Assays Enzyme Assays
BACE2 (β-Secretase-2, Memapsin-1) belongs to the family of transmembrane aspartic proteases
The SensoLyte® 520 BACE2 Assay Kit is a homogeneous assay that can be used to detect the activity of enzyme and for screening of BACE2 inhibitors. It contains a QXL® 520/ HiLyte™ Fluor488 FRET substrate, derived from a peptide sequence surrounding cleavage site of BACE2. QXL® 520 quenches HiLyte™ Fluor488 fluorescence in the intact FRET peptide. Active BACE2 cleaves FRET substrate into two separate fragments resulting in the release of HiLyte™ Fluor488 fluorescence that can be monitored at excitation /emission= 490/520 nm. The long wavelength fluorescence of HiLyte™ Fluor488 is less interfered by the autofluorescence of components in biological samples and test compounds.
BACE2 has approximately 75% homology with BACE1 (β-Secretase-1) and is similar in that both cleaves Amyloid Precursor Protein (APP) at β-site generating N-terminal part of beta-Amyloid peptide. Unlike BACE1 that is highly expressed in pancreas and brain tissue, BACE2 is more widespread and can be found in many peripheral tissues at higher levels compared to BACE1. BACE2 is one of the highest efficiency beta-amyloid peptide degrading enzyme that is less active only to insulin degrading enzyme (IDE); therefore, targeting BACE2 is considered a potential therapeutic strategy for the prevention and treatment of Alzheimer's disease.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty