Would you like to log in again?
SensoLyte® 520 ADAM10 Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72226
103010-668EA 887.15 USD InStock
103010-668
SensoLyte® 520 ADAM10 Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
ADAM10 has been shown to be an effective α-secretase in vitro and in vivo
The SensoLyte® 520 ADAM10 Activity Assay Kit is an optimized assay that can be used to detect the ADAM 10 activity. The unique FRET substrate contained in the kit was designed to reduce the cross reactivity with ADAM17 (also called tumor necrosis factor- α-converting enzyme, TACE). When active ADAM10 cleaves the FRET substrate, it results in an increase of 5-FAM fluorescence monitored at excitation/emission = 490/520 nm. The long wavelength fluorescence of 5-FAM is also less interfered by the autofluorescence of components in biological samples and test compounds. This assay can detect as low as 0.25 ng/mL active ADAM10.
This enzyme acts as a sheddase to cleave cell surface proteins involved in neuropathology, inflammatory response and tumor progression.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty