Would you like to log in again?
SensoLyte® 520 TEV Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72227
103010-670EA 700.08 USD InStock
103010-670
SensoLyte® 520 TEV Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Tobacco Etch Virus (TEV) proteinase is widely used to remove fusion tags from recombinant proteins
The SensoLyte® TEV Protease Assay Kit uses TEV specific Glu-Asn-Leu-Tyr-Phe-Gln-Gly substrate labeled with 5-FAM dye and QXL® 520 quencher. Proteolytic cleavage of this quenched 5-FAM-QXL® 520 conjugate yields a brightly green fluorescence, which can be continuously monitored at Ex/Em= 490/520 nm. Increase in fluorescence intensity is directly proportional to the TEV protease quantity/activity.
It recognizes specific sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly and cleaves between Glutamine and Glycine residues. TEV protease is active even at 4 °C, making it the protease of choice for fusion proteins or tags removal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty