Would you like to log in again?
SensoLyte® Green Glutaminyl Cyclase Activity Assay (Fluorimetric), AnaSpec Inc.
Supplier: AnaSpec
AS-72230
103010-676EA 1043.54 USD InStock
103010-676
SensoLyte® Green Glutaminyl Cyclase Activity Assay (Fluorimetric), AnaSpec Inc.
Assays Enzyme Assays
The SensoLyte® Green Glutaminyl Cyclase Activity Assay Kit provides a convenient, two-step homogeneous procedure for measuring enzyme activity from various sources using a green fluorescence substrate
Many biologically active peptides and proteins have pyroglutamate (pGlu) as their first amino acid. This modification protects them from degradation by amino peptidases and in many cases determines functionality of protein or peptide. However, pyroglutamate modification sometimes may lead to negative side effects. For example, pGlu modified beta-amyloid peptides (pGlu-Aβ) aggregate much faster compared to non-pyroglutamic ones and form toxic oligomers that may damage neurons. Recent studies linked pGlu-Aβ to the Alzheimer’s Disease because it was identified as a major component of amyloid plaques. pGlu can be formed when glutamine or glutamic acid spontaneously convert into pyroglutamate. However, cyclic reaction rate is dramatically increased in the presence of Glutaminyl Cyclase (QC), also known as Glutaminyl-peptide Cyclotransferase (QPCT). Human Glutaminyl Cyclase has been isolated from nerve tissue, and detected in cerebrospinal fluid and saliva.
During the first step, substrate is incubated with Glutaminyl Cyclase or enzyme-containing samples and converted into the pyroglutamate form. Secondly, glutaminyl cyclase developer is added to remove pGlu residue and generate the green fluorophore. It can be detected with excitation at 490 nm and emission at 520 nm. Produced fluorescence is proportional to the enzyme activity. This kit is ideal for high throughput screening (HTS) of glutaminyl cyclase activators and inhibitors. The long wavelength of the green fluorophore is less interfered by autofluorescence of cell components and test compounds. Assay sensitivity is 1.5 ng/mL of active enzyme.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty