Would you like to log in again?
SensoLyte® 520 IDE Activity Assay Kit (Fluorimetric), AnaSpec
Supplier: AnaSpec
AS-72231
103010-678EA 931 USD InStock
103010-678
SensoLyte® 520 IDE Activity Assay Kit (Fluorimetric), AnaSpec
Assays Enzyme Assays
Insulin degrading enzyme (IDE) is considered a physiological and pathological relevant enzyme
The SensoLyte® 520 IDE Activity Assay Kit can be used to detect enzyme activity in biological samples or in purified enzyme preparations. The unique FRET substrate was derived from an APP sequence designed to reduce cross reactivity with Neprilysin, ADAM10, TACE, BACE-1, and BACE-2. When active IDE cleaves the FRET substrate, it results in an increase of 5-FAM fluorescence, which is monitored at excitation/emission = 490 nm/520 nm. The long wavelength fluorescence of 5-FAM is also less interfered by the autofluorescence of components in biological samples and test compounds. This assay can detect as low as 0.8 ng/mL active IDE.
Growing evidence has indicated that IDE may be involved in the pathogenesis of Alzheimer’s disease and diabetes mellitus type 2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty