Would you like to log in again?
Aminoallyl dUTP, 4 mM in TE buffer, UltraPure Grade, Anaspec
Supplier: AnaSpec
AS-83203
103011-126EA 298.15 USD InStock
103011-126
Aminoallyl dUTP, 4 mM in TE buffer, UltraPure Grade, Anaspec
Nucleic Acid Reagents Nucleotides
Building block for preparing amino-containing nucleotide.
Caution: Store at -20°C, desiccated and protected from light
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty