Would you like to log in again?
CAPILLARY THETA GLASS PREMIUM PK100
Supplier: HARVARD APPARATUS MS
Theta Glass ^ Theta Glass, Premium Capillary, Model TG150-4, Dielectric Constant: 4.6, Glass Composition: 81% SiO2, 13% B2O3, 4% Na2O, 2% Al2O3, Glass Type: Corning 7740 Borosilicate (Pyrex), ID Metric: 1.00, Length Metric: 100, OD Metric: 1.5, Septum Metric: 0.2
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPILLARY THETA GLASS PREMIUM PK100 103011-534 64-0810 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty