Would you like to log in again?
pH Standard Buffers, Highly Acidic, Inorganic Ventures
Supplier: Inorganic Ventures
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PH-3-10L PH-1.68-4L PH-1.68-1L PH-2-500ML PH-1.68-250ML PH-3-1L PH-1.68-500ML PH-2-250ML PH-2-4L PH-2-1L PH-3-4L PH-3-250ML PH-3-500ML PH-2-10L
103050-872EA 128.99 USD InStock
103050-872 103050-860 103050-882 103050-862 103050-884 103050-870 103050-880 103050-858 103050-868 103050-876 103050-864 103050-886 103050-878 103050-866
pH Standard Buffers, Highly Acidic, Inorganic Ventures
Buffers pH Calibration Buffers
  • Certified reference material set in a water matrix for stability.
  • These standards are Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines.
  • One year Expiration Date.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty