Would you like to log in again?
pH Standard Buffers, Inorganic Ventures
Supplier: Inorganic Ventures
PHRED-4-10L PH-10-10L PH-6-10L PH-9.18-250ML PH-6-250ML PH-8-500ML PHBLUE-10-1L PHRED-4-1L PH-4-500ML PH-9.18-500ML PHBLUE-10-4L PH-5-1L PH-10-500ML PH-8-250ML PH-6-4L PHBLUE-10-10L PH-5-500ML PH-4-1L PH-8-4L PH-4-4L PHRED-4-250ML PH-9-500ML PH-6-1L PHBLUE-10-500ML PH-9-10L PH-10-4L PH-8-1L PH-4-250ML PHRED-4-500ML PH-10-1L PH-9-1L PH-6-500ML PH-4-10L PH-9.18-1L PH-5-4L PH-9-250ML PHRED-4-4L PH-5-250ML PH-8-10L PH-9-4L PHBLUE-10-250ML
103051-016EA 42.77 USD InStock
103051-016 103050-894 103050-992 103050-970 103050-972 103050-994 103050-890 103050-990 103050-946 103050-968 103050-924 103050-926 103050-964 103050-920 103050-942 103050-986 103050-988 103050-944 103050-966 103050-922 103050-888 103051-000 103050-928 103050-982 103050-984 103050-962 103050-940 103050-980 103050-912 103050-978 103050-914 103050-930 103050-996 103050-974 103050-976 103050-998 103050-932 103050-938 103051-014 103050-916 103050-918
pH Standard Buffers, Inorganic Ventures
Buffers pH Calibration Buffers
  • Certified reference material set in a water matrix for stability.
  • These standards are Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines.
  • One year Expiration Date.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty