Would you like to log in again?
PIPETTE TRNSR GRAD FINE 104MM 1ML PK400
Supplier: SARSTEDT INC MS
Transfer pipette 1ml^ Transfer pipette 1 ml, without graduation, Length: 104 mm, Drop size: 17 - 22 ul, Max. Volume 1 ml, Sample volume 1 ml, material: Low Density Polyethylene (LD-PE), Colour: Transparent, No CE marking
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PIPETTE TRNSR GRAD FINE 104MM 1ML PK400 103061-606 86.1180 Pack of 400 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty