Would you like to log in again?
BOWL W/FOOD/WATER CONTAINERS FOR ANIMALS
Supplier: 0000013487
CMA 120 Bowl^ CMA 120 Bowl, with Food and Water Containers, for Freely Moving Animals enables Microdialysis studies on conscious, small laboratory animals over long periods of time. The additions of these containers allow the animal to feed and drink ad libitum.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOWL W/FOOD/WATER CONTAINERS FOR ANIMALS 103133-554 CMA8309672 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty