Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C1orf69 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-15075R-FITC
10316-544EA 792.2 USD InStock
10316-544
Anti-C1orf69 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Belonging to the CAF17 subfamily of the gcvT family, C1orf69 is a 356 amino acid mitochondrial protein that is required for normal heme synthesis. Heme synthesis is dependent upon receiving iron through iron sulfur (Fe-S) cluster biogenesis proteins such as C1orf69, which is specifically thought to assemble Fe-S clusters for mitochondrial aconitase and lipoate synthase. In zebrafish, deficiency of C1orf69 causes anemia. The gene encoding C1orf69 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1.

Type: Primary
Antigen: C1orf69
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 131-230/256
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty