Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Quick-Load Purple Low Molecular Weight DNA Ladder, New England Biolabs
Supplier: New England Biolabs
N0557S N0557S
103218-932EA 100 USD InStock
CA103218-932 103218-932
Quick-Load Purple Low Molecular Weight DNA Ladder, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
Quick-Load Purple Low Molecular Weight DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye.
  • Convenient, ready-to-load format with NEB's proprietary purple loading dye
  • Size range: 25 to 766 bp
  • Sharp bands
  • No UV Shadow
  • Easy to identify reference bands
Tracking dye does not leave a shadow under UV light. The DNA Ladder consists of a proprietary plasmid which is digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The 200 base pair band has increased intensity to serve as a reference point.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty