Would you like to log in again?
BsrFI-v2, New England Biolabs
Supplier: New England Biolabs
R0682S R0682S
103218-954EA 75 USD InStock
CA103218-954 103218-954
BsrFI-v2, New England Biolabs
Media for Microbiology Additives and Supplements for Microbiology
A restriction endonuclease that recognizes the sequence R^CCGG_Y.
  • Sourced from an E.coli strain that carries the cloned and modified BsrFI gene
  • Cleavage of mammalian genomic DNA is blocked by CpG methylation
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty