Would you like to log in again?
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
A10 A10 A10BX
10324-332PK 198.38 USD InStock
CA89405-024 10324-332 89405-024
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.
  • Optimized for superior performance
  • For the non-selective cultivation of microorganisms. General growth medium for bacteria, yeast, and fungi.
  • Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals.
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in sleeves of 10 plates, in ""breathable"" cellulose bags that prevents build-up of condensation and excess moisture.
  • The 100 plate size box offers superior protection during shipment and is easy to open without a cutting tool.
For the non-selective cultivation of microorganisms. General growth media for bacteria, yeast, and fungi. Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals. Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding. Packaged in sleeves of 10 plates in a "breathable" bag that prevents build-up of condensation and excess moisture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty