Would you like to log in again?
LJ (Lowenstein Jensen) Media, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
X19 X19 C25 X22
10324-414PK 163.87 USD InStock
10324-414 10324-412 10324-368 CA10001-506
LJ (Lowenstein Jensen) Media, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the selective isolation of Mycobacterium spp.
  • Selective isolation, cultivation
For the selective isolation and cultivation of Mycobacterium spp.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty