Would you like to log in again?
Tryptic Soy Broth (TSB) with 15% Glycerol, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
D09 D09 D08 D08
10789-404PK 485.5 USD InStock
10789-404 CA10789-404 10324-374 CA10001-418
Tryptic Soy Broth (TSB) with 15% Glycerol, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A cryopreservative medium for the freezing of cultures.
  • Freeze cultures
  • Long-term preservation
A cryopreservative medium for the freezing of cultures for long term preservation.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty