Would you like to log in again?
Mueller Hinton Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
R38
10324-388PK 60.66 USD InStock
10324-388
Mueller Hinton Broth, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the preparation of suspensions for disk diffusion sensitivity testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty