Would you like to log in again?
AlbiQuick™ Candida Albicans Differentiation, Five Minute Test, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z121 Z121
10324-428PK 286.85 USD InStock
10324-428 CA10001-514
AlbiQuick™ Candida Albicans Differentiation, Five Minute Test, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
The new Hardy AlbiQuick™ utilizes two enzyme color reactions to produce an accurate result that’s easy to read out.
  • Test for Candida albicans in only five minutes
  • Performed on a convenient card with two reaction wells
  • Eliminates lengthy procedures and tedious microscopy
Now you can test for Candida albicans in only five minutes! The new Hardy AlbiQuick™ utilizes two enzyme color reactions to produce an accurate result that’s easy to read out
Requires the use of a handheld 366nm ultra violet lamp.
The old germ tube method of C. albicans identification requires several hours of incubation and tedious microscopy to produce a result. The AlbiQuick™ eliminates the need for this and other lengthy procedures.
A recent study, which compared the above NAG and PRO methods to the traditional germ tube test, showed a sensitivity of 99% and a specificity of 99-100%.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty