Would you like to log in again?
NALC Powder for Sputum Decontamination and Digestion, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Z60 Z60
10324-432PK 148.8 USD InStock
CA10001-526 10324-432
NALC Powder for Sputum Decontamination and Digestion, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Used to break down mucous components of sputum and other clinical specimens.
  • Allow slower growing mycobacteria to grow
  • Decontaminate normal flora specimens
  • Break down mucous components of sputum
Used to break down mucous components of sputum and other clinical specimens and to decontaminate the specimen of normal flora in order to allow slower growing mycobacteria to grow. Use with TB Base Digestant.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty