Would you like to log in again?
Brucella Agar/Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
D04 J87
10324-448PK 32.53 USD InStock
10324-448 10068-016
Brucella Agar/Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of anaerobic bacteria and the selective isolation of Bacteroides spp.
  • Broth for preservation of organisms by freezing
  • Cultivation of anaerobic bacteria
  • Selective isolation of Bacteroides
For the cultivation of anaerobic bacteria and the selective isolation of Bacteroides spp. Brucella Broth with 15% Glycerol for the preservation of organisms by freezing, packaged in a glass cryovial with a black screw-cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty