Would you like to log in again?
Rapid Ornithine, for Enterobacteriaceae and Staphylococcus Lugdunensis
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K279 K279
10324-464PK 106.42 USD InStock
10324-464 CA10001-470
Rapid Ornithine, for Enterobacteriaceae and Staphylococcus Lugdunensis
Media for Microbiology Dehydrated Media for Microbiology
This test will assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis.
  • Detect ornithine decarboxylase activity in as little as two to four hours
  • Assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis
Hardy Diagnostics Rapid Ornithine can be used to detect ornithine decarboxylase activity in as little as two to four hours. This test will assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty