Would you like to log in again?
MycoVue™, Slide Culture System, Potato Flake Agar, Ready-To-Use, Hardy Diagnostics
Supplier: Hardy Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MV1
10324-474BX 266.14 USD InStock
10324-474
MycoVue™, Slide Culture System, Potato Flake Agar, Ready-To-Use, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
A ready-to-use slide culture system for the cultivation of fungi.
  • Simplify your fungal slide cultures while saving money
  • Culture can be incubated for long periods of time
  • Easy observation without disturbing the fungal structures
A ready-to-use slide culture system for the cultivation of fungi. Simplify your fungal slide cultures while saving money, reducing frustration, and eliminating preparation time. Desiccation is prevented with a built in humidity chamber and the culture can be incubated for long periods of time. Device fits conveniently on the microscope stage for easy observation without disturbing the fungal structures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty