Would you like to log in again?
GBS Detect/Strep Reagents, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z175 G123 Z122 G123 Z75 A300 GA300 Z75 A70
10324-504PK 80.78 USD InStock
10324-504 CA10001-426 10324-436 CA10001-530 10053-788 10324-430 10324-342 10324-334 76288-444
GBS Detect/Strep Reagents, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.
  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping, includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™, to aid in the identification of gram positive cocci
GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty