Would you like to log in again?
Blood Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
GA10 J32BX J32 J22 H28 J93 GA10
10324-546PK 374.81 USD InStock
10324-546 75860-384 10324-522 10324-544 10158-957 CA10001-438 10324-542
Blood Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Hardy Diagnostics Blood Agar products are recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.
  • Microorganism isolation, cultivation, differentiation
  • General purpose growth media
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty