Would you like to log in again?
Haemophilus ID Quadplate, for X and V Factors, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J82 J82
10324-556PK 87.02 USD InStock
CA10001-458 10324-556
Haemophilus ID Quadplate, for X and V Factors, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Haemophilus ID Quadplate consists of four separate formulations
  • RTF Casman Medium
  • Modified (X- and V-factors)
  • Tryptic Soy Agar (TSA)
  • Chocolate Agar (X- and V-factors)
Haemophilus ID Quadplate consists of four separate formulations, which supply the requirements as follows: RTF Casman Medium, Modified (X- and V-factors); Tryptic Soy Agar (TSA) with Hemin (X-factor); Tryptic Soy Agar (TSA), modified with NAD (V-factor) ; Chocolate Agar (X- and V-factors).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty