Would you like to log in again?
Rose Agar/MacConkey Agar Biplate, Hardy Diagnostics
Supplier: Hardy Diagnostics
J85 J85
10324-558PK 31.36 USD InStock
10324-558 CA10001-460
Rose Agar/MacConkey Agar Biplate, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Rose Agar/MacConkey Agar Biplate is used for gram positive/negative bacteria.
  • Rose Agar for gram positive bacteria
  • MacConkey Agar for gram negative bacteria
Rose Agar, selective for gram positive bacteria. MacConkey Agar, selective for gram negative bacteria.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty