Would you like to log in again?
Germ Tube Cryo™ for Candida Albicans, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z217
10324-568PK 165.56 USD InStock
10324-568
Germ Tube Cryo™ for Candida Albicans, Hardy Diagnostics
Cell Culture Supplements and Additives
A ready-to-use germ tube solution made up of stabilized animal serum.
  • Comes pre-measured in vials
  • No mixing, measuring, or aliquoting is required
  • Eliminates exposure to human serum
A ready-to-use germ tube solution made up of stabilized animal serum, comes pre-measured in vials. No mixing, measuring, or aliquoting is required! Eliminates exposure to human serum. 0.5ml in each vial. Germ tubes develop on C. albicans within one hour. Serum formula is enhanced for more rapid and luxuriant production of germ tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty