Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PYR Test Kit, with 15 ml Chromogenic Solution and Test Cards, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z275 Z275
10324-574EA 278.25 USD InStock
CA10001-522 10324-574
PYR Test Kit, with 15 ml Chromogenic Solution and Test Cards, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
PYR Test Kit and PYR Reagent are recommended for the detection of pyrolidonyl arylamidase activity.
  • Three tests can be performed on one card
  • Card format is easier to handle than disks
Hardy Diagnostics PYR Test Kit and PYR Reagent are recommended for the detection of pyrolidonyl arylamidase (also called pyrolidonyl aminopeptidase) activity in certain groups of bacteria, such as Streptococcus pyogenes (group A strep), Enterococcus spp., some coagulase-negative staphylococci, and some Enterobacteriaceae. The card format is easier to handle than disks. Three tests can be performed on one card.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty