Would you like to log in again?
WesternBright MCF Western Blotting Kit, Advansta
Supplier: Advansta
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K-12021-010 K-12022-010 K-12023-010
103254-712EA 269.79 USD InStock
103254-712 103254-714 103254-716
WesternBright MCF Western Blotting Kit, Advansta
Blotting Kits
Multi-color fluorescent Western blotting in one convenient kit.
  • Multiplex - visualize two proteins at once
  • With pg detection, 10x more sensitive than Cy dyes
  • Only 3.5 hrs for entire protocol
  • Includes quality Western blotting reagents from Advansta
  • Compatible with Cy3/5 detecting or near-IR systems
  • Choose between visible (APC/RPE) or near-IR (IR700/800) kits
WesternBright MCF visible and near infrared (IR) fluorescent Western blotting kits allow the assay of two proteins at once, increasing the quality and quantity of information that can be gained from a single blot.
Ordering information: Kit includes secondary antibodies, blocking solution, washing solution, pre-cut PVDF membranes, background quenching sheets.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty