Would you like to log in again?
AdvanWash™ washing solution, Advansta
Supplier: Advansta
R-03024-D50 R-03100-D50
103254-750EA 190.38 USD InStock
103254-750 103254-752
AdvanWash™ washing solution, Advansta
Buffers Electrophoresis Buffers Washing Buffers
Optimized washing solutions, for Western blotting with WesternBright systems.
  • Reliable and quality controlled for reproducible results
  • Compatible with chemiluminescent and fluorescent Western blots
  • Saves time by providing a pre-made buffer without mixing or adjusting pH
AdvanWash and AdvanWash-IR washing solutions are optimized to work with Advansta Western blotting system. Choose AdvanWash for all WesternBright chemiluminescent HRP substrates and the WesternBright MCF fluorescent Western blotting kit, and AdvanWash-IR for the WesternBright MCF-IR kits.
Provided as a 10X concentrate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty