Would you like to log in again?
AdvanStain Scarlet, Advansta
Supplier: Advansta
AdvanStain Scarlet™
K-11072-C25 K-11072-B50
103254-918EA 237.69 USD InStock
103254-918 103254-796
AdvanStain Scarlet, Advansta
Stains Electrophoresis Stains
Fluorescent protein stain for gels and blots.
  • Sensitive enough to detect less than nanogram of protein
  • Convenient and fast results in 3 hours
  • Flexible for staining gels or membranes
  • Reversible to destain for downstream Western blotting or mass spectrometry
  • No speckling in the background for cleaner data
  • Biodegradable with no heavy metals for increased safety and simple disposal
  • Compatible with any laser or CCD imaging system - green (543, 532nm), blue (488nm), violet (405nm), and UV (302, 365nm), with maximum of 610 nm
AdvanStain Scarlet is a fluorescent protein stain for gels and blots that allows sensitive and quantitative visualization of protein bands. With a quick and easy protocol, gels can be stained in less than 3 hours, and imaged on a wide variety of imaging systems. Gels and blots stained with AdvanStain Scarlet can be imaged on any fluorescence imaging system, including laser- and CCD-based systems, with a detection limit of less than 1 ng of protein per band or spot. Fully compatible with downstream Western blotting or mass spectrometry, AdvanStain Scarlet is non-toxic and biodegradable, for safe and simple disposal.
Ordering information: Includes AdvanStain Scarlet Dye concentrate, Powder A for fix solution, and Powder B for stain buffer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty