Would you like to log in again?
ROTOR PKG MLS-50 SWNG BKT 50000RPM ALM
Supplier: BECKMAN COULTER LIFE SCIENCES, MS
MLS-50 Rotor Package 4 x 5.0 mL^ MLS-50 Rotor Package, Ultra - Swinging Bucket, Material: Aluminum, Volume: 4 x 5.0 mL, Maximum Speed: 50,000 rpm, Maximum RCF at r max (g): 268,000 x g, Applications: Rate-zonal separation of subcellular organelles, pelleting and isopycnic separations of RNA
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROTOR PKG MLS-50 SWNG BKT 50000RPM ALM 103255-122 367280 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty