Would you like to log in again?
ROTOR TLS-55 AY 55000RPM ALM 4X2.2ML
Supplier: BECKMAN COULTER LIFE SCIENCES, MS
TLS-55 Rotor Swinging Bucket^ TLS-55 Rotor, Ultra - Swinging Bucket, Maximum Speed: 55,000 rpm, Maximum RCF at r max (g): 259,000 x g, k-Factor: 50, Rotor Material: Anodized Aluminum, Bucket Material: Titanium, Anodized Aluminum Cap, No. Of Rotor Cavities x Nominal Largest Load Capacity: 4 x 2.2 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ROTOR TLS-55 AY 55000RPM ALM 4X2.2ML 103255-156 346936 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty