Would you like to log in again?
Protein Deglycosylation Mix II, New England Biolabs
Supplier: New England Biolabs
P6044S P6044S
103257-624EA 586 USD InStock
CA103257-624 103257-624
Protein Deglycosylation Mix II, New England Biolabs
Protein Purification Protein Purification Reagents
The Protein Deglycosylation Mix II contains all of the enzymes, reagents, and controls needed to remove all N-linked and simple O-linked glycans as well as some complex O-linked glycans.
  • Fast reaction setup
  • Complete sugar removal with no protein degradation
  • Recombinant enzymes with no contaminating endoglycosidase or exoglycosidases
  • Each enzyme exhibits ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
This mix contains enzyme sufficient for 20 reactions or the cleavage of as much as 2 mg of glycoprotein. All of the enzymes and reagents included in the Protein Deglycosyation Mix II are Mass Spectrometry compatible. Following the deglycosylation reaction, samples are ready to be prepared for mass spectrometry analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty