Would you like to log in again?
MagID Magnetic Isolation Device, OZ Biosciences
Supplier: OZ Biosciences
DM30000
103258-106EA 136.45 USD InStock
103258-106
MagID Magnetic Isolation Device, OZ Biosciences
Transfection Reagents
OZ Biosciences MagID Magnetic Isolation Device is ideal for coupling reactions, washing procedures, aspiration, pipetting etc.
  • Ideal for your magnetic nanoparticles coupling reaction and purification
  • Adapted to working solutions ranging from 10 µL to 2 mL
  • Perfect for a quick magnetic separation process (< 5 minutes) with a high yield separation
  • Durable and easy to use with an open faced design
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty