Would you like to log in again?
Lullaby™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Lullaby™
LL71000 LL70500 LL73000
103258-116EA 428.89 USD InStock
103258-116 103258-114 103258-118
Lullaby™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Lullaby is the ideal siRNA transfection reagent for gene silencing
  • Effective with very low doses of siRNA
  • Minimized off-target effects
  • Suitable for siRNA, shRNA, dsRNA, etc.
  • Applicable to a broad range of cells
  • Serum compatible & non-toxic
  • Straightforward protocol
Relying on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and a very low toxicity.
Delivery information: Shipped at room temperatue
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty