Would you like to log in again?
ViroMag™ R/L Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
ViroMag™
RL40100 KC30700 RL41000 RL40200
103258-142EA 1221.92 USD InStock
103258-142 103258-138 103258-418 103258-416
ViroMag™ R/L Transduction Reagent, OZ Biosciences
Transfection Reagents
ViroMag R/L Transduction Reagent is a magnetic nanoparticles formulation optimized for Retroviruses and Lentiviruses.
  • Increase transduction efficiency
  • Accelerate the transduction process
  • Efficient for hard-to-infect and non-permissive cells
  • Concentrate the viral dose onto the cells
  • Suitable for all type of Retro/Lentivirus
  • Synchronise cells adsorption/infection without modifications of the viruses
Based on the Magnetofection™ technology, this reagent allows concentrating the complete applied dose of Retro/Lentiviral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty