Would you like to log in again?
AdenoMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
AdenoMag™
AM71000 AM70100 AM70200 KC30900
103258-140EA 924 USD InStock
103258-140 103258-146 103258-144 103258-148
AdenoMag™ Transfection Reagent, OZ Biosciences
Transfection Reagents
AdenoMag Transfection Reagent is a magnetic nanoparticles formulation specifically dedicated to enhance Adenovirus and Adeno Associated Virus (AAV) infection and transduction capacities.
  • Increase transduction efficiency
  • Accelerate the transduction process
  • Efficient for hard-to-infect and non-permissive cells
  • Concentrate the viral dose onto the cells
  • Synchronise cells adsorption/infection without modifications of the viruses
Based on the Magnetofection™ technology, it allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty