Would you like to log in again?
In vivo ViroMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
ViroMag™
IV-KC30230 IV-VM30250 IV-VM30500 IV-TK30230
103258-152EA 472.51 USD InStock
103258-152 103258-284 103258-274 103258-286
In vivo ViroMag™ Transfection Reagent, OZ Biosciences
Transfection Reagents
In vivo ViroMag Transfection Reagent has been designed for in vivo targeted transduction with any viral vector
  • High and increased transduction/infection efficiency
  • Magnetically targeted transduction to specific area
  • Reduction of virus dose and systemic dissemination
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the vector into tissues
  • Minimized toxicity
  • Suitable for all types of viruses
Virus/nanoparticles complexes can be easily administrated through various injection routes such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store at +4 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty