Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D(-)-Luciferin sodium salt ≥99.5% endotoxin-free

Supplier: OZ Biosciences
Synonyms: D-Luciferin, sodium4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt

LN10000
103258-194EA 590.02 USD InStock
103258-194
D(-)-Luciferin sodium salt ≥99.5% endotoxin-free
D(-)-Luciferin sodium salt
OZ Biosciences D-Luciferin K+ and Na+ salts are routinely used as Firefly’s Luciferase substrate in in vitro and in vivo bioluminescent assays. The quality and purity of the D-Luciferin are essential to obtain good and reproducible results. OZ Biosciences is offering high quality of Endotoxin-Free D-Luciferin K+ and Na+ salts.
Store at -20°C and protect from light.

  • High purity > 99.5%
  • Good solubility and great sensitivity
  • Reliable in vivo reporter for bioluminescent assays
  • Endotoxin free (ideal for in vivo application)
  • Quick and easy diffusion throughout the animal

Suitable for bioluminescent assays, whole animal imaging, ATP assays, immunoassays, pyrosequencing, luciferase fragment complementation for sequential gene analysis experiments.

Delivery information: Shipped at room temperature


Formula: C₁₁H₇N₂NaO₃S₂
MW: 302.30 g/mol
MDL Number: MFCD00044938
CAS Number: 103404-75-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty