Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade

Supplier: OZ Biosciences
Synonyms: BCIG(2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

XG31000
103258-198EA 132.72 USD InStock
103258-198
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal substrate is metabolized by the β-galactosidase enzyme into an insoluble blue precipitate. It is ideal for staining transformed bacteria and LacZ transfected or infected cells, tissues and organisms.

  • High Purity >99%
  • Molecular biology grade
  • Great sensitivity
  • Perfect for cells, tissues and organisms staining

The quality and purity of the X-Gal substrate is essential to obtain high-quality and reliable results.

Delivery information: Shipped at room temperature.

Caution: Store at –20 °C.


Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty