Would you like to log in again?
AlumVax Hydroxide, OZ Biosciences
Supplier: OZ Biosciences
AH0050 AH0250
103258-200EA 138.46 USD InStock
103258-200 103258-332
AlumVax Hydroxide, OZ Biosciences
Protein Purification Protein Purification Resins
AlumVax Hydroxide is wet gel (colloidal) of aluminum hydroxide 2%, provided as a ready-to-use suspension
Aluminum hydroxide is the most common adjuvant used in approved prophylactic vaccines because of its excellent safety profile and ability to enhance protective humoral immune response. Since more than 80 years, it has been observed that aluminium compounds act by a depot effect and also by direct activation of the immune cells. Adsorption or entrapment of antigens in aggregates through hydrophobic and electrostatic interactions favors a high local antigen concentration and improved uptake by antigen presentating cells (APC).
AlumVax hydroxide is a crystalline aluminum oxyhydroxide that is positively charged at physiological pH (pI=11), suitable for adsorption of negatively charged, acidic proteins (such as albumin).
Delivery information: Shipped at room temperature
Packaging: Store at room temperature
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty